Skip to Content
Merck
Periodic acid 99.999% trace metals basis

Periodic acid

Periodic acid 99.999% trace metals basis
Synonym(s):
Periodic acid
Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reaction Type
Reagent Type
Special Grade
Technique