![Peretinoin ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/204/384/879c7ad3-02e5-41b8-86f3-69bb5b209f5d/640/879c7ad3-02e5-41b8-86f3-69bb5b209f5d.png)
Peretinoin
![Peretinoin ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/204/384/879c7ad3-02e5-41b8-86f3-69bb5b209f5d/640/879c7ad3-02e5-41b8-86f3-69bb5b209f5d.png)
Synonym(s):
3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic acid
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing