Skip to Content
Merck
Peptide YY porcine ≥97% (HPLC), synthetic

Peptide YY porcine

Peptide YY porcine ≥97% (HPLC), synthetic
Synonym(s):
PYY, Tyr-Pro-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2
Empirical Formula (Hill Notation):
C190H288N54O57
CAS Number:
Molecular Weight:
4240.65

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing