![Pentoxyverine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/690/801/1f229374-55a2-4a99-8614-4ae9be52457c/640/1f229374-55a2-4a99-8614-4ae9be52457c.png)
Pentoxyverine impurity A
![Pentoxyverine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/690/801/1f229374-55a2-4a99-8614-4ae9be52457c/640/1f229374-55a2-4a99-8614-4ae9be52457c.png)
Synonym(s):
1-Phenyl-1-cyclopentanecarboxylic acid
Linear Formula:
C6H5C5H8CO2H
CAS Number:
Molecular Weight:
190.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing