Skip to Content
Merck
Pentachlorobenzene certified reference material (for chromatographic applications), suitable for qNMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Pentachlorobenzene

Pentachlorobenzene certified reference material (for chromatographic applications), suitable for qNMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Linear Formula:
HC6Cl5
CAS Number:
Molecular Weight:
250.34
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique