![Pelargonin chloride ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/314/897/0ce053d1-2764-4e14-9eac-392df67d9953/640/0ce053d1-2764-4e14-9eac-392df67d9953.png)
Pelargonin chloride
![Pelargonin chloride ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/314/897/0ce053d1-2764-4e14-9eac-392df67d9953/640/0ce053d1-2764-4e14-9eac-392df67d9953.png)
Synonym(s):
3,5-Bis(glucosyloxy)-4′,7-dihydroxyflavylium chloride, Monardin, Pelargonidin 3,5-di-O-glucoside chloride
Empirical Formula (Hill Notation):
C27H31ClO15
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing