Skip to Content
Merck
PEGA resin extent of labeling: ~0.4 mmol/g loading

PEGA resin

PEGA resin extent of labeling: ~0.4 mmol/g loading
Synonym(s):
Poly[acryloyl-bis(aminopropyl)polyethylene glycol]

Products

Product Category
Brand
Physical Form
Product Line
Functional Group
Particle Size (µm)
Chain Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing