Skip to Content
Merck
PCP-D5 (Phencyclidine-D5) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

PCP-D5 (Phencyclidine-D5) solution

PCP-D5 (Phencyclidine-D5) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H20ND5
CAS Number:
Molecular Weight:
248.42

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique