Skip to Content
Merck
p-Anisaldehyde primary reference standard

p-Anisaldehyde

p-Anisaldehyde primary reference standard
Synonym(s):
4-Methoxybenzaldehyde, Aubépine
Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing