
Pancuronium bromide

Synonym(s):
1,1′-[(2β,3α,5α,16β,17β)-3,17-bis(Acetyloxy)androstane-2,16-diyl]bis(1-methylpiperidinium) dibromide
Empirical Formula (Hill Notation):
C35H60Br2N2O4
CAS Number:
Molecular Weight:
732.67
Products
Product Category
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3