Palmitic acid-d31
Synonym(s):
Hexadecanoic-d31 acid
Linear Formula:
CD3(CD2)14CO2H
CAS Number:
Molecular Weight:
287.62
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing