Skip to Content
Merck
Paeoniflorin European Pharmacopoeia (EP) Reference Standard

Paeoniflorin

Paeoniflorin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
NSC 178886, Peoniflorin
Empirical Formula (Hill Notation):
C23H28O11
CAS Number:
Molecular Weight:
480.46
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4