![Paclitaxel natural for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/389/274/b0ed0f40-c837-4c1f-85a1-9f7851f0995a/640/b0ed0f40-c837-4c1f-85a1-9f7851f0995a.png)
Paclitaxel natural for peak identification
![Paclitaxel natural for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/389/274/b0ed0f40-c837-4c1f-85a1-9f7851f0995a/640/b0ed0f40-c837-4c1f-85a1-9f7851f0995a.png)
Synonym(s):
Paclitaxel
Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing