
Oxfendazole

Synonym(s):
Methyl 5-(phenylsulfinyl)-benzimidazol-2-carbamate
Empirical Formula (Hill Notation):
C15H13N3O3S
CAS Number:
Molecular Weight:
315.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing