Skip to Content
Merck
Orysastrobin PESTANAL®, analytical standard

Orysastrobin

Orysastrobin PESTANAL®, analytical standard
Synonym(s):
(2E)-2-(Methoxyimino)-2-{2-[(3E,5E,6E)-5-(methoxyimino)-4,6-dimethyl-2,8-dioxa-3,7-diazanona-3,6-dien-1-yl]phenyl}-N-methylacetamide
Empirical Formula (Hill Notation):
C18H25N5O5
CAS Number:
Molecular Weight:
391.42

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing