Ondansetron Related Compound C
Synonym(s):
9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, 1,2,3,9-Tetrahydro-9-methyl-4H-carbazol-4-one
Empirical Formula (Hill Notation):
C13H13NO
CAS Number:
Molecular Weight:
199.25
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing