
Omeprazole magnesium

Empirical Formula (Hill Notation):
C34H36MgN6O6S2
CAS Number:
Molecular Weight:
713.12
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing