Ofloxacin Related Compound A
Synonym(s):
(3 RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid)
Empirical Formula (Hill Notation):
C17H18FN3O4
CAS Number:
Molecular Weight:
347.34
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing