![N-Sulfocarbamoylgonyautoxin-2 and N-Sulfocarbamoylgonyautoxin-3 solution 20 μg/g in acetic acid (nominal concentration)](/deepweb/assets/sigmaaldrich/product/images/336/998/a670091a-57ae-4731-8006-6a94dc32df19/640/a670091a-57ae-4731-8006-6a94dc32df19.jpg)
N-Sulfocarbamoylgonyautoxin-2 and N-Sulfocarbamoylgonyautoxin-3 solution
![N-Sulfocarbamoylgonyautoxin-2 and N-Sulfocarbamoylgonyautoxin-3 solution 20 μg/g in acetic acid (nominal concentration)](/deepweb/assets/sigmaaldrich/product/images/336/998/a670091a-57ae-4731-8006-6a94dc32df19/640/a670091a-57ae-4731-8006-6a94dc32df19.jpg)
Synonym(s):
Toxin C1,2
Empirical Formula (Hill Notation):
C10H17N7O11S2
Molecular Weight:
475.41
Products
Brand
Formula Weight
Format
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing