Skip to Content
Merck
N-Octanoyl-N-methylglucamine ≥97% (GC)

N-Octanoyl-N-methylglucamine

N-Octanoyl-N-methylglucamine ≥97% (GC)
Synonym(s):
N-(D-Glucityl)-N-methyloctanamide, N-Methyl-N-octanoyl-D-glucamine, MEGA-8, OMEGA
Empirical Formula (Hill Notation):
C15H31NO6
CAS Number:
Molecular Weight:
321.41

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing