Skip to Content
Merck
Nocodazole ≥99% (TLC), powder

Nocodazole

Nocodazole ≥99% (TLC), powder
Synonym(s):
Methyl N-(5-thenoyl-2-benzimidazolyl)carbamate, Methyl [5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]carbamate, Oncodazole, R 17934, [5-(2-Thienylcarbonyl)-1H-benzimidazol-2-yl]-carbamic acid methyl ester, Methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate, Nocidazole, Nocodazolum, InSolution Nocodazole
Empirical Formula (Hill Notation):
C14H11N3O3S
CAS Number:
Molecular Weight:
301.32

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment