![Nilotinib Related compound C](/deepweb/assets/sigmaaldrich/product/structures/350/061/c8386485-3195-4e29-a16c-a4975639c074/640/c8386485-3195-4e29-a16c-a4975639c074.png)
Nilotinib Related compound C
![Nilotinib Related compound C](/deepweb/assets/sigmaaldrich/product/structures/350/061/c8386485-3195-4e29-a16c-a4975639c074/640/c8386485-3195-4e29-a16c-a4975639c074.png)
Synonym(s):
3-Amino-4-methylbenzoic acid, 3-Amino-p-toluic acid
Linear Formula:
H2NC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
151.16
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing