
Nicosulfuron

Synonym(s):
1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(dimethylcarbamoyl)-2-pyridylsulfonyl]urea, 2-[(4,6-Dimethoxypyrimidin-2-ylcarbamoyl)sulfamoyl]-N,N-dimethylnicotinamide
Empirical Formula (Hill Notation):
C15H18N6O6S
CAS Number:
Molecular Weight:
410.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2