Skip to Content
Merck
N-Ethylbenzylamine 97%

N-Ethylbenzylamine

N-Ethylbenzylamine 97%
Synonym(s):
N-Benzylethylamine
Linear Formula:
C6H5CH2NHCH2CH3
CAS Number:
Molecular Weight:
135.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing