![Nerolidol 98%, Mixture of cis and trans](/deepweb/assets/sigmaaldrich/product/structures/384/184/0e1cebf5-15f2-461f-950e-49517321cce7/640/0e1cebf5-15f2-461f-950e-49517321cce7.png)
Nerolidol
![Nerolidol 98%, Mixture of cis and trans](/deepweb/assets/sigmaaldrich/product/structures/384/184/0e1cebf5-15f2-461f-950e-49517321cce7/640/0e1cebf5-15f2-461f-950e-49517321cce7.png)
Synonym(s):
3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol
Linear Formula:
(CH3)2C[=CHCH2CH2C(CH3)]2(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3