Skip to Content
Merck
Neocuproine hydrochloride monohydrate for spectrophotometric det. of Cu, ≥99.0%

Neocuproine hydrochloride monohydrate

Neocuproine hydrochloride monohydrate for spectrophotometric det. of Cu, ≥99.0%
Synonym(s):
2,9-Dimethyl-1,10-phenanthroline hydrochloride
Empirical Formula (Hill Notation):
C14H12N2 · HCl · H2O
CAS Number:
Molecular Weight:
262.73

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1