N-Benzylquininium chloride
Synonym(s):
QUIBEC
Empirical Formula (Hill Notation):
C27H31ClN2O2
CAS Number:
Molecular Weight:
451.00
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing