Narirutin
Synonym(s):
Narirutin, (2S)-7-((6-O-(6-Deoxy-a-L-mannopyranosyl)-β-D-glucopyranosyl)oxy)-2,3-dihydro-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Isonaringenin, Isonaringin, Naringenin rutinoside
Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing