
Naproxen Related Compound L

Synonym(s):
6′-Methoxy-2′-acetonaphthone, 1-(6-Methoxy-2-naphthalenyl)ethanone, 2-Acetyl-6-methoxynaphthalene, NSC 105564
Linear Formula:
CH3OC10H6COCH3
CAS Number:
Molecular Weight:
200.23
Products
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing