![Naphazoline Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/373/962/0876cb87-5c2e-4302-a617-4c3c5612cc28/640/0876cb87-5c2e-4302-a617-4c3c5612cc28.jpg)
Naphazoline Related Compound A
![Naphazoline Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/373/962/0876cb87-5c2e-4302-a617-4c3c5612cc28/640/0876cb87-5c2e-4302-a617-4c3c5612cc28.jpg)
Synonym(s):
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide
Empirical Formula (Hill Notation):
C14H16N2O
CAS Number:
Molecular Weight:
228.29
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing