
Naphazoline hydrochloride

Synonym(s):
2-(1-Naphthylmethyl)imidazoline hydrochloride, Naphazoline hydrochloride
Empirical Formula (Hill Notation):
C14H14N2 · HCl
CAS Number:
Molecular Weight:
246.74
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3