
N-Acetylglucosamine

Synonym(s):
N-Acetyl-D-glucosamine, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing