N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
Synonym(s):
2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine, N,N-Bis(trifluoromethylsulfonyl)-5-chloro-2-pyridylamine, Comins’ Reagent
Empirical Formula (Hill Notation):
C7H3ClF6N2O4S2
CAS Number:
Molecular Weight:
392.68
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing