Myristic acid-1-13C
Synonym(s):
Tetradecanoic acid-1-13C
Linear Formula:
CH3(CH2)1213CO2H
CAS Number:
Molecular Weight:
229.36
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing