Mupirocin
Synonym(s):
5,9-Anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-[2E,8[2S,3S(1S,2S)]]-L-talonon-2-enonic acid 8-carboxyoctyl ester, BRL 4910A, Pseudomonic acid, Mupirocin
Empirical Formula (Hill Notation):
C26H44O9
CAS Number:
Molecular Weight:
500.62
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Agency
Research Area
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing