Monoclonal Anti-TYRP1 antibody produced in mouse
Synonym(s):
CAS2, TRP, TYRP, TYRRP
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing