![Monoclonal Anti-Cytokeratin Peptide 18 antibody produced in mouse clone KS-B17.2, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/149/873/7da74827-c0c5-4311-aebd-005745668cdd/640/7da74827-c0c5-4311-aebd-005745668cdd.jpg)
Monoclonal Anti-Cytokeratin Peptide 18 antibody produced in mouse
![Monoclonal Anti-Cytokeratin Peptide 18 antibody produced in mouse clone KS-B17.2, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/149/873/7da74827-c0c5-4311-aebd-005745668cdd/640/7da74827-c0c5-4311-aebd-005745668cdd.jpg)
Synonym(s):
Monoclonal Anti-Cytokeratin Peptide 18
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2