Skip to Content
Merck
Mevalonolactone-(methyl-d3) 99 atom % D, ≥97% (CP)

Mevalonolactone-(methyl-d3)

Mevalonolactone-(methyl-d3) 99 atom % D, ≥97% (CP)
Synonym(s):
DL-Mevalonic-methyl-d3 lactone
Empirical Formula (Hill Notation):
C6D3H7O3
Molecular Weight:
133.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing