Metosulam
Synonym(s):
N-(2,6-Dichloro-3-methylphenyl)-5,7-dimethoxy-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide
Empirical Formula (Hill Notation):
C14H13Cl2N5O4S
CAS Number:
Molecular Weight:
418.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2