Metoclopramide Related Compound D
Synonym(s):
Methyl 4-acetamido-2-methoxybenzoate
Empirical Formula (Hill Notation):
C11H13NO4
CAS Number:
Molecular Weight:
223.23
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing