Skip to Content
Merck
Methyl Tricosanoate certified reference material, pharmaceutical secondary standard

Methyl tricosanoate

Methyl Tricosanoate certified reference material, pharmaceutical secondary standard
Synonym(s):
Tricosanoic acid methyl ester, Methyl tricosanoate
Linear Formula:
CH3(CH2)21COOCH3
CAS Number:
Molecular Weight:
368.64

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique