
Methyl palmitoleate

Synonym(s):
Methyl cis-9-hexadecenoate, Palmitoleic acid methyl ester
Linear Formula:
CH3(CH2)5CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
268.43
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Tissue
Quality Segment
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3