Skip to Content
Merck
Methyl furfuryl disulfide ≥95%, FG

Methyl furfuryl disulfide

Methyl furfuryl disulfide ≥95%, FG
Synonym(s):
Furfuryl methyl disulfide
Empirical Formula (Hill Notation):
C6H8OS2
CAS Number:
Molecular Weight:
160.26
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing