Skip to Content
Merck
Methyl Benzylidene Camphor Pharmaceutical Secondary Standard; Certified Reference Material

Methyl Benzylidene Camphor

Methyl Benzylidene Camphor Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-(4-Methylbenzylidene)camphor, 1,7,7-Trimethyl-3-(4-methylbenzylidene)-norbornan-2-one, 1,7,7-Trimethyl-3-[(4-methylphenyl)methylene]-bicyclo[2.2.1]heptan-2-one, 4-MBC, 4-Methylbenzylidene camphor
Empirical Formula (Hill Notation):
C18H22O
CAS Number:
Molecular Weight:
254.37

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1