Methyl arachidonate
Synonym(s):
Arachidonic acid methyl ester
Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing