![Methyl 3-[[(2RS)-2-bromopropanoyl]amino]-4-methylthiophene-2-carboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/331/768/fbeabdee-e24a-485c-9d26-3232f0427df1/640/fbeabdee-e24a-485c-9d26-3232f0427df1.png)
Methyl 3-[[(2RS)-2-bromopropanoyl]amino]-4-methylthiophene-2-carboxylate
![Methyl 3-[[(2RS)-2-bromopropanoyl]amino]-4-methylthiophene-2-carboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/331/768/fbeabdee-e24a-485c-9d26-3232f0427df1/640/fbeabdee-e24a-485c-9d26-3232f0427df1.png)
Empirical Formula (Hill Notation):
C10H12BrNO3S
CAS Number:
Molecular Weight:
306.18
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1