Skip to Content
Merck
(Methoxymethyl)triphenylphosphonium chloride 97%

(Methoxymethyl)triphenylphosphonium chloride

(Methoxymethyl)triphenylphosphonium chloride 97%
Synonym(s):
Methoxymethyl-triphenylphosphonium chloride
Linear Formula:
CH3OCH2P(C6H5)3Cl
CAS Number:
Molecular Weight:
342.80

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2