Skip to Content
Merck
(Methoxymethyl)trimethylsilane 98%

(Methoxymethyl)trimethylsilane

(Methoxymethyl)trimethylsilane 98%
Synonym(s):
(Trimethylsilyl)methyl methyl ether
Linear Formula:
CH3OCH2Si(CH3)3
CAS Number:
Molecular Weight:
118.25

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing