Skip to Content
MilliporeSigma
Methaqualone-D7 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Methaqualone-D7 solution

Methaqualone-D7 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C16H7D7N2O
CAS Number:
Molecular Weight:
257.34
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing