![Methacrylic acid - methylmethacrylate copolymer (1:1) European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/100/061/89a9c1b2-6b59-4409-b23c-2579d48ecd7d/640/89a9c1b2-6b59-4409-b23c-2579d48ecd7d.png)
Methacrylic acid - methylmethacrylate copolymer (1:1)
![Methacrylic acid - methylmethacrylate copolymer (1:1) European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/100/061/89a9c1b2-6b59-4409-b23c-2579d48ecd7d/640/89a9c1b2-6b59-4409-b23c-2579d48ecd7d.png)
Synonym(s):
Poly(methyl methacrylate-co-methacrylic acid), Poly(methacrylic acid-co-methyl methacrylate)
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing